missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cystatin SA Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
£339.08 - £816.39
Specifications
Assay Range | 2.4 pg/mL |
---|---|
Assay Sensitivity | 2.34-150 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18288927
|
Novus Biologicals™
NBP2-79652-15PLATES |
15 Plates |
£816.39
Pack of 15 |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18220976
|
Novus Biologicals™
NBP2-79652-5PLATES |
5 Plates |
£339.08
Pack of 5 |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cystatin SA.Specifications
2.4 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cystatin SA. | |
Sandwich ELISA | |
1470 | |
Mouse | |
RUO | |
Cystatin SA |
2.34-150 pg/mL | |
HRP | |
Antibody Pairs | |
Human | |
cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
CST2 | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 150 pg/mL is recommended. |