Learn More
Cytiva Cytodex 1 Gamma Microcarriers
Microcarrier for general use in sterilized dry powder form.
Brand: Cytiva 17548701
Additional Details : Weight : 0.03000kg
Description
- Cytodex 1 Gamma is designed by substituting a cross-linked dextran matrix with positively charged N, N-diethylaminoethyl (DEAE) groups distributed throughout the matrix.
- Micro-Carriers Dry Weight: 4.3 × 106
- Surface Area Dry Weight: 0.44 m2/g
Specifications
Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, diff erentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving celltransfer and studies on uptage of labelled compounds | |
30 g |