missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Cytodex 1 Gamma Microcarriers
Microcarrier for general use in sterilized dry powder form.
£1027.00 - £32000.00
Specifications
For Use With (Application) | Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, diff erentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving celltransfer and studies on uptage of labelled compounds |
---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15727207
|
Cytiva
17548702 |
300 g |
£4721.00
300g |
Estimated Shipment: 22-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15717207
|
Cytiva
17548701 |
30 g |
£1027.00
30g |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15737207
|
Cytiva
17548703 |
3 kg |
£32000.00
3kg |
Estimated Shipment: 22-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Cytodex 1 Gamma is designed by substituting a cross-linked dextran matrix with positively charged N, N-diethylaminoethyl (DEAE) groups distributed throughout the matrix.
- Micro-Carriers Dry Weight: 4.3 × 106
- Surface Area Dry Weight: 0.44 m2/g
Specifications
Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, diff erentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving celltransfer and studies on uptage of labelled compounds |