missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

£32.70 - £310.00

Chemical Identifiers

CAS 14316-06-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00066141
InChI Key IYUKFAFDFHZKPI-UHFFFAOYNA-N
Synonym d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885
PubChem CID 11182647
IUPAC Name methyl (2R)-2-aminopropanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(C)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15418988
View Documents
Thermo Scientific Alfa Aesar
H62472.06
5 g
£32.70
5g
Please to purchase this item. Need a web account? Register with us today!
15428988
View Documents
Thermo Scientific Alfa Aesar
H62472.14
25 g
£108.00
25g
Please to purchase this item. Need a web account? Register with us today!
15438988
View Documents
Thermo Scientific Alfa Aesar
H62472.22
100 g
£310.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.

Solubility
Soluble in Water 100 mg/ml.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

14316-06-4
139.58
IYUKFAFDFHZKPI-UHFFFAOYNA-N
11182647
[H+].[Cl-].COC(=O)C(C)N
C4H10ClNO2
MFCD00066141
d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885
methyl (2R)-2-aminopropanoate;hydrochloride

Specifications

108°C to 110°C
White
MFCD00066141
d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885
IYUKFAFDFHZKPI-UHFFFAOYNA-N
methyl (2R)-2-aminopropanoate;hydrochloride
11182647
98%
D-Alanine methyl ester hydrochloride
14316-06-4
C4H10ClNO2
4235975
Soluble in Water 100mg/ml.
[H+].[Cl-].COC(=O)C(C)N
139.58
139.58
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 238-258-3

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado