missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ D-Biotin
D-Biotin
Brand: Molecular Probes™ B1595
94.05 GBP valid until 2024-12-31
Use promo code "21979" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Description
D-biotin binds with high affinity to streptavidin, avidin and NeutrAvidin™ biotin-binding proteins. This solution can be used in conjunction with unlabeled streptavidin or avidin in procedures to block endogenous biotin.Specifications
Unconjugated | |
Store at room temperature. | |
Solid | |
Biotin (unconjugated) |
1 g | |
Room Temperature | |
Invitrogen™ |
For Research Use Only. Not for use in diagnostic procedures.