missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ D-Biotin
D-Biotin
£103.00 - £115.00
Specifications
Conjugate | Unconjugated |
---|---|
Shipping Condition | Room Temperature |
Product Line | Invitrogen™ |
Product Type | Biotin (unconjugated) |
Description
D-biotin binds with high affinity to streptavidin, avidin and NeutrAvidin™ biotin-binding proteins. This solution can be used in conjunction with unlabeled streptavidin or avidin in procedures to block endogenous biotin.Specifications
Unconjugated | |
Room Temperature | |
Invitrogen™ | |
Biotin (unconjugated) |
For Research Use Only. Not for use in diagnostic procedures.