Learn More
Molecular Probes™ D-Biotin
D-Biotin
Brand: Molecular Probes™ B20656
76.67 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Description
D-biotin binds with high affinity to streptavidin, avidin and NeutrAvidin™ biotin-binding proteins. This solution can be used in conjunction with unlabeled streptavidin or avidin in procedures to block endogenous biotin.Specifications
50mM | |
10 mL | |
Room Temperature | |
Invitrogen™ |
Unconjugated | |
Store in refrigerator (2–8°C). | |
Liquid | |
Biotin (unconjugated) |
Safety and Handling
- N-(2,6-Diethylphenyl)maleimide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : N-(2,6-Diethylphenyl)maleimide
For Research Use Only. Not for use in diagnostic procedures.