missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(-)-Fructose, Certified AR for Analysis, Fisher Chemical™
£92.05 - £274.00
Chemical Identifiers
CAS | 57-48-7 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.156 |
MDL Number | 148910 |
InChI Key | BJHIKXHVCXFQLS-UYFOZJQFSA-N |
Synonym | d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300 |
PubChem CID | 5984 |
ChEBI | CHEBI:48095 |
IUPAC Name | (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
SMILES | C(C(C(C(C(=O)CO)O)O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10111620
|
Fisher Chemical
F/1952/48 |
100 g |
£92.05
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10488210
|
Fisher Chemical
F/1952/53 |
500 g |
£148.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10579760
|
Fisher Chemical
F/1952/60 |
1 kg |
£274.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D(-)-Fructose, Certified AR for Analysis
Chemical Identifiers
57-48-7 | |
180.156 | |
BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
5984 | |
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
C6H12O6 | |
148910 | |
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300 | |
CHEBI:48095 | |
C(C(C(C(C(=O)CO)O)O)O)O |
Specifications
57-48-7 | |
White | |
Plastic powder jar | |
148910 | |
BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one | |
5984 | |
180.16g/mol |
103°C | |
5 to 7 | |
C6H12O6 | |
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300 | |
C(C(C(C(C(=O)CO)O)O)O)O | |
180.156 | |
CHEBI:48095 | |
Solid |
Safety and Handling
Product Identifier
- D(-)-Fructose
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title