missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucono-1,5-lactone, 99%, Thermo Scientific Chemicals

 

£20.50 - £101.51

Chemical Identifiers

CAS 90-80-2
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00006647
InChI Key PHOQVHQSTUBQQK-UHFFFAOYNA-N
Synonym gluconolactone, delta-gluconolactone, d-glucono-1,5-lactone, d-gluconolactone, 1,5-gluconolactone, d-gluconic acid lactone, gluconic acid lactone, glucono delta-lactone, 1,5-d-gluconolactone, gluconic lactone
PubChem CID 7027
ChEBI CHEBI:16217
SMILES OCC1OC(=O)C(O)C(O)C1O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11344529
View Documents
Thermo Scientific Alfa Aesar
A13105.30
250 g
£20.50
250g
Please to purchase this item. Need a web account? Register with us today!
11494444
View Documents
Thermo Scientific Alfa Aesar
A13105.36
500 g
£26.33
500g
Please to purchase this item. Need a web account? Register with us today!
Currently Unavailable: We have no information when this product will be available
View alternative products
11404454
View Documents
Thermo Scientific Alfa Aesar
A13105.0E
2500 g
£101.51
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glucono delta-lactone (GDL) is a kind of excellent multifunctional food additive. It is mainly used as protein coagulant, acidifier, pickling, or leavening agent. expander, preservative, seasonings, sequestrant, chelating agent, color-preserver, etc. it is widely applied to food, daily chemical industry, pharmaceuticals, cosmetics. Plastic and resinous modification, electrode position and polish, metal-surface cleaning and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glucono delta-lactone (GDL) is a kind of excellent multifunctional food additive. It is mainly used as protein coagulant, acidifier, pickling, or leavening agent. expander, preservative, seasonings, sequestrant, chelating agent, color-preserver, etc. it is widely applied to food, daily chemical industry, pharmaceuticals, cosmetics. Plastic and resinous modification, electrode position and polish, metal-surface cleaning and organic synthesis.

Solubility
Soluble in water 500 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep awy from strong oxidizing agents, strong bases.
Specifications

Chemical Identifiers

90-80-2
178.14
PHOQVHQSTUBQQK-UHFFFAOYNA-N
7027
OCC1OC(=O)C(O)C(O)C1O
C6H10O6
MFCD00006647
gluconolactone, delta-gluconolactone, d-glucono-1,5-lactone, d-gluconolactone, 1,5-gluconolactone, d-gluconic acid lactone, gluconic acid lactone, glucono delta-lactone, 1,5-d-gluconolactone, gluconic lactone
CHEBI:16217

Specifications

90-80-2
C6H10O6
83286
gluconolactone, delta-gluconolactone, d-glucono-1,5-lactone, d-gluconolactone, 1,5-gluconolactone, d-gluconic acid lactone, gluconic acid lactone, glucono delta-lactone, 1,5-d-gluconolactone, gluconic lactone
PHOQVHQSTUBQQK-UHFFFAOYNA-N
OCC1OC(=O)C(O)C(O)C1O
178.14
CHEBI:16217
99%
154°C to 159°C (decomposition)
MFCD00006647
14,4457
Soluble in water 500g/L (20°C).
+65° (c=4 in Water)
(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
7027
178.14
D-(+)-Glucono-1,5-lactone
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 202-016-5

missing translation for 'rtecsNumber' : LZ5184000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.