missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals β-D-Glucose pentaacetate, 98%
£16.40 - £214.07
Chemical Identifiers
CAS | 604-69-3 |
---|---|
MDL Number | MFCD00006597 |
InChI Key | LPTITAGPBXDDGR-IBEHDNSVSA-N |
Synonym | beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-β |
PubChem CID | 2724702 |
IUPAC Name | [(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate |
SMILES | CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10495423
|
Thermo Scientific Chemicals
302960050 |
5 g | Glass Bottle | N/A | |||||
10013460
|
Thermo Scientific Chemicals
302961000 |
100 g | Plastic bottle |
£56.98
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10096193
|
Thermo Scientific Chemicals
302965000 |
500 g | Plastic bottle |
£214.07
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
604-69-3 | |
LPTITAGPBXDDGR-IBEHDNSVSA-N | |
2724702 | |
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C |
MFCD00006597 | |
beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-β | |
[(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate |
Specifications
604-69-3 | |
100.0 | |
Authentic | |
C16H22O11 | |
+ 5.00 (20.00°C c=1,CHCL3) | |
beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-β | |
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C | |
390.341 | |
390.33 | |
Crystalline Powder |
97.5 | |
White | |
98% | |
MFCD00006597 | |
+4° to +6° (20°C, 589nm) (c=1, CHCl3) | |
LPTITAGPBXDDGR-IBEHDNSVSA-N | |
[(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate | |
2724702 | |
≥97.5% | |
β-D-Glucose pentaacetate |
RUO – Research Use Only