missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronic acid, 98%, Thermo Scientific Chemicals

£74.37 - £363.81

Chemical Identifiers

CAS 6556-12-3
Molecular Formula C6H10O7
Molecular Weight (g/mol) 194.14
MDL Number MFCD00064368
InChI Key IAJILQKETJEXLJ-QTBDOELSSA-N
ChEBI CHEBI:47953
IUPAC Name (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10747783
View Documents
Thermo Scientific Acros
309880100
10 g Plastic bottle
£74.37
10g
Please to purchase this item. Need a web account? Register with us today!
10472071
View Documents Promotion Details
Thermo Scientific Acros
309880250
25 g Plastic bottle
£142.71
25g
Please to purchase this item. Need a web account? Register with us today!
10531984
View Documents
Thermo Scientific Acros
309881000
100 g Plastic bottle
£363.81
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6556-12-3
194.14
IAJILQKETJEXLJ-QTBDOELSSA-N
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
C6H10O7
MFCD00064368
CHEBI:47953

Specifications

6556-12-3
White
98%
MFCD00064368
+ 36.50 (20.00°C c=6,H2O,after 2 hrs)
2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
C(=O)C(C(C(C(C(=O)O)O)O)O)O
194.14
CHEBI:47953
98%
D-Glucuronic acid, 98%
158.0°C to 162.0°C
Authentic
C6H10O7
15,4501
+ 36.50
IAJILQKETJEXLJ-QTBDOELSSA-N
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
65041
194.14
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'einecsNumber' : 229-486-4

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only