Learn More
Tocris Bioscience™ D-Luciferin sodium salt
Firefly luciferase substrate
Brand: Tocris Bioscience™ 5427/10
64.79 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
D-Luciferin sodium salt is a firefly luciferase substrate (Km = ∽2 μM). Used in many standard bioluminescence imaging (BLI) techniques, including monitoring tumour growth, and investigating intracellular signaling activity in vitro and in vivo. Cell permeable. Emission maximum = 562 nm.Specifications
>99% | |
10 mg | |
302.3 |
C11H7N2NaO3S2 | |
D-Luciferin sodium salt | |
D-Luciferin sodium salt |
For Research Use Only