missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ D-Luciferin sodium salt

Firefly luciferase substrate

£85.00 - £254.00

Specifications

Purity >99%
Molecular Formula C11H7N2NaO3S2
Product Type D-Luciferin sodium salt
Molecular Weight (g/mol) 302.3
Inhibitors D-Luciferin sodium salt
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18743988
View Documents
Tocris Bioscience™
5427/50
50 mg
£254.00
50mg
Please to purchase this item. Need a web account? Register with us today!
18706009
View Documents Promotion Details
Tocris Bioscience™
5427/10
10 mg
£85.00
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Firefly luciferase substrate. Cell permeable.
Specifications

Specifications

>99%
D-Luciferin sodium salt
D-Luciferin sodium salt
C11H7N2NaO3S2
302.3
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only