Learn More
Thermo Scientific Chemicals D-Mannitol, 97+%
£23.70 - £187.00
Chemical Identifiers
CAS | 69-65-8 |
---|---|
Molecular Formula | C6H14O6 |
Molecular Weight (g/mol) | 182.17 |
MDL Number | MFCD00064287 |
InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
Synonym | d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex |
PubChem CID | 6251 |
ChEBI | CHEBI:16899 |
IUPAC Name | (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol |
SMILES | OCC(O)C(O)C(O)C(O)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11437916
|
Thermo Scientific Chemicals
A14030.30 |
250 g |
£23.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427916
|
Thermo Scientific Chemicals
A14030.0B |
1000 g |
£45.90
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11447916
|
Thermo Scientific Chemicals
A14030.0I |
5000 g |
£187.00
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-Mannitol is used in the determination of glomerular filtration rate. It is used as an inert osmotic control substance. Further, it is used in the treatment of oliguric renal failure. It is also used as a sweetener for people with diabetes, in chewing gums and acts as an adulterant for heroin, methamphetamines and cocaine. It finds application as a research tool in cell biological studies, usually to control osmolarity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Mannitol is used in the determination of glomerular filtration rate. It is used as an inert osmotic control substance. Further, it is used in the treatment of oliguric renal failure. It is also used as a sweetener for people with diabetes, in chewing gums and acts as an adulterant for heroin, methamphetamines and cocaine. It finds application as a research tool in cell biological studies, usually to control osmolarity.
Solubility
Soluble in water, alcohol, aniline, pyridine and glycerol. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
69-65-8 | |
182.17 | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
6251 | |
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol |
C6H14O6 | |
MFCD00064287 | |
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
CHEBI:16899 | |
OCC(O)C(O)C(O)C(O)CO |
Specifications
69-65-8 | |
1.489 | |
Odorless | |
MFCD00064287 | |
14,5745 | |
Soluble in water,alcohol,aniline,pyridine and glycerol. Insoluble in ether. | |
+24° (in Borax Solution) | |
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol | |
6251 | |
182.17 | |
D-Mannitol |
165°C to 167°C | |
290°C to 295°C (3.5 mmHg) | |
C6H14O6 | |
1721898 | |
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
OCC(O)C(O)C(O)C(O)CO | |
182.17 | |
CHEBI:16899 | |
97+% |
Safety and Handling
missing translation for 'einecsNumber' : 200-711-8
missing translation for 'rtecsNumber' : OP2060000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only