missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Mannitol, 97+% Product Code.: 11437916

Thermo Scientific Chemicals D-Mannitol, 97+%

Product Code. 11437916
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11437916

Brand: Thermo Scientific Chemicals A14030.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-Mannitol is used in the determination of glomerular filtration rate. It is used as an inert osmotic control substance. Further, it is used in the treatment of oliguric renal failure. It is also used as a sweetener for people with diabetes, in chewing gums and acts as an adulterant for heroin, methamphetamines and cocaine. It finds application as a research tool in cell biological studies, usually to control osmolarity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Mannitol is used in the determination of glomerular filtration rate. It is used as an inert osmotic control substance. Further, it is used in the treatment of oliguric renal failure. It is also used as a sweetener for people with diabetes, in chewing gums and acts as an adulterant for heroin, methamphetamines and cocaine. It finds application as a research tool in cell biological studies, usually to control osmolarity.

Solubility
Soluble in water, alcohol, aniline, pyridine and glycerol. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 165°C to 167°C
Density 1.489
Boiling Point 290°C to 295°C (3.5 mmHg)
Odor Odorless
Quantity 250 g
Beilstein 1721898
Merck Index 14,5745
Solubility Information Soluble in water,alcohol,aniline,pyridine and glycerol. Insoluble in ether.
Optical Rotation +24° (in Borax Solution)
Formula Weight 182.17
Percent Purity 97+%
Chemical Name or Material D-Mannitol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251092 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14030.0I,A14030.30,A14030.0B
Lot Number10251752 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14030.0I,A14030.30,A14030.0B
Lot Number10250633 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14030.0I,A14030.30,A14030.0B
Lot Number10245272 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog NumberA14030.0I,A14030.30,A14030.0B
Lot Number10243972 Certificate TypeCertificate of Analysis Date04/11/2022 Catalog NumberA14030.0I,A14030.30,A14030.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Mannitol, 97+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.