missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Mannitol, ACS reagent, Thermo Scientific Chemicals
£33.17 - £360.80
Chemical Identifiers
CAS | 69-65-8 |
---|---|
Molecular Formula | C6H14O6 |
Molecular Weight (g/mol) | 182.17 |
MDL Number | MFCD00064287 |
InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
Synonym | d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex |
PubChem CID | 6251 |
ChEBI | CHEBI:16899 |
IUPAC Name | hexane-1,2,3,4,5,6-hexol |
SMILES | OCC(O)C(O)C(O)C(O)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10269650
|
Thermo Scientific Acros
423922500 |
250 g | Plastic bottle |
£33.17
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10148230
|
Thermo Scientific Acros
423920010 |
1 kg | Plastic bottle |
£88.34
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10358673
|
Thermo Scientific Acros
423920050 |
5 kg | Plastic drum |
£360.80
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69-65-8 | |
182.17 | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
6251 | |
hexane-1,2,3,4,5,6-hexol |
C6H14O6 | |
MFCD00064287 | |
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
CHEBI:16899 | |
OCC(O)C(O)C(O)C(O)CO |
Specifications
69-65-8 | |
White | |
0.05% max. (5 g, 105°C, 2 hrs) | |
C6H14O6 | |
01, IV, 2841 | |
+23.3° to +24.3° (25°C, 589nm) (Reagent ACS-directives) | |
Solubility in water: soluble. Other solubilities: soluble in pyridine and aniline,slightly soluble in lower alcohols,practically insoluble in most other common,organic solvents | |
OCC(O)C(O)C(O)C(O)CO | |
182.17 | |
CHEBI:16899 | |
ACS Reagent | |
0.0008 meq/g max | |
D-Mannitol |
165.0°C to 170.0°C | |
0.01% max. | |
Authentic | |
MFCD00064287 | |
15, 5810 | |
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
hexane-1,2,3,4,5,6-hexol | |
6251 | |
182.17 | |
0.01% max | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 200-711-8
RUO – Research Use Only