missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(-)-Quinic acid, 98+%, Thermo Scientific Chemicals
£39.00 - £518.00
Chemical Identifiers
CAS | 77-95-2 |
---|---|
Molecular Formula | C7H12O6 |
Molecular Weight (g/mol) | 192.17 |
MDL Number | MFCD00003864 |
InChI Key | AAWZDTNXLSGCEK-UHFFFAOYNA-N |
Synonym | quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi |
PubChem CID | 6508 |
IUPAC Name | (3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid |
SMILES | OC1CC(O)(CC(O)C1O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10146130
|
Thermo Scientific Acros
160500250 |
25 g | Glass bottle |
£39.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10659014
|
Thermo Scientific Acros
160501000 |
100 g | Glass bottle |
£108.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11995071
|
Thermo Scientific Acros
160505000 |
500 g | Glass bottle |
£518.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
77-95-2 | |
192.17 | |
AAWZDTNXLSGCEK-UHFFFAOYNA-N | |
6508 | |
OC1CC(O)(CC(O)C1O)C(O)=O |
C7H12O6 | |
MFCD00003864 | |
quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi | |
(3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid |
Specifications
77-95-2 | |
White to Yellow | |
2% max. | |
98+% | |
(HO)4C6H7CO2H | |
15,8175 | |
− 44.00 | |
Solubility in water: 400g/L (20°C). Other solubilities: soluble in glacial acetic acid and alcohol | |
OC1CC(O)(CC(O)C1O)C(O)=O | |
192.17 | |
192.17 | |
Crystalline Powder |
166.0°C to 168.0°C | |
0.1% max. | |
Authentic | |
C7H12O6 | |
MFCD00003864 | |
- 44.00 (20.00°C c=11.2,H2O) | |
quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi | |
AAWZDTNXLSGCEK-UHFFFAOYNA-N | |
(3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid | |
6508 | |
98+% | |
D(-)-Quinic acid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-072-8
RUO – Research Use Only