missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Sorbitol, 97%
£20.50 - £217.08
Chemical Identifiers
CAS | 50-70-4 |
---|---|
Molecular Formula | C6H14O6 |
Molecular Weight (g/mol) | 182.17 |
MDL Number | MFCD00004708 |
InChI Key | FBPFZTCFMRRESA-JGWLITMVSA-N |
Synonym | d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol |
PubChem CID | 5780 |
ChEBI | CHEBI:17924 |
IUPAC Name | (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol |
SMILES | C(C(C(C(C(CO)O)O)O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10616830
|
Thermo Scientific Chemicals
132730050 |
5 g | Glass bottle |
£21.71
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10813781
|
Thermo Scientific Chemicals
132731000 |
100 g | Plastic bottle |
£20.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10418870
|
Thermo Scientific Chemicals
132735000 |
500 g | Plastic bottle |
£31.56
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10396820
|
Thermo Scientific Chemicals
132730010 |
1 kg | Plastic bottle |
£48.44
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10113570
|
Thermo Scientific Chemicals
132730025 |
2.5 kg | Plastic bottle |
£95.37
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10185253
|
Thermo Scientific Chemicals
132730100 |
10 kg | Plastic drum |
£217.08
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
50-70-4 | |
182.17 | |
FBPFZTCFMRRESA-JGWLITMVSA-N | |
5780 | |
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol |
C6H14O6 | |
MFCD00004708 | |
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol | |
CHEBI:17924 | |
C(C(C(C(C(CO)O)O)O)O)O |
Specifications
50-70-4 | |
White | |
96% min. (on anhydrous substance) (HPLC) | |
MFCD00004708 | |
158851 | |
+ 4.00 | |
Solubility in water: soluble. Other solubilities: slightly soluble in acetic acid, soluble in glycerol and propylene glycol, slightly soluble in methanol, ethanol, phenol,, acetamide, acetone, dmf and pyridine, practically insoluble in most other org. sol | |
C(C(C(C(C(CO)O)O)O)O)O | |
182.17 | |
CHEBI:17924 | |
97% | |
D-Sorbitol |
95.0°C to 99.0°C | |
Authentic | |
C6H14O6 | |
1533 | |
+ 4.00 (20.00°C per eur. pharm.) | |
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol | |
FBPFZTCFMRRESA-JGWLITMVSA-N | |
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol | |
5780 | |
182.17 | |
Granular Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 200-061-5
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals D-Sorbitol, 97%