missing translation for 'onlineSavingsMsg'
Learn More
D-(-)-Tartaric acid, 99%, Thermo Scientific Chemicals Product Code.: 11473614

D-(-)-Tartaric acid, 99%, Thermo Scientific Chemicals

Product Code. 11473614
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11473614

Brand: Thermo Scientific Alfa Aesar A11264.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.

Solubility
Soluble in water and alcohol.

Notes
Light sensitive. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-71-7
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00004238
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acidShow MoreShow Less
PubChem CID 439655
ChEBI CHEBI:15672
IUPAC Name (2S,3S)-2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

pH 1.6
Melting Point 166°C to 170°C
Beilstein 1725145
Merck Index 14,9068
Solubility Information Soluble in water and alcohol.
Formula Weight 150.09
Percent Purity 99%
Quantity 25 g
Flash Point 210°C (410°F)
Odor Odorless
Optical Rotation −13° (c=10 in water)
Chemical Name or Material D- (-)-Tartaric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62500098 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberA11264.22,A11264.36,A11264.14
Lot Number10253179 Certificate TypeCertificate of Analysis Date29/11/2024 Catalog NumberA11264.22,A11264.14,A11264.36
Lot NumberZ04J014 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog NumberA11264.22,A11264.14,A11264.36
Lot NumberM24J010 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog NumberA11264.22,A11264.14,A11264.36
Lot NumberM19I028 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11264.22,A11264.14,A11264.36
compliance-icons
Product Identifier
  • D-(-)-Tartaric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : D-(-)-Tartaric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-(-)-Tartaric acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.