missing translation for 'onlineSavingsMsg'
Learn More

D-tert-Leucine, 99%, Thermo Scientific Chemicals

£23.20 - £224.00

Chemical Identifiers

CAS 26782-71-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00004265
InChI Key NPDBDJFLKKQMCM-UHFFFAOYNA-N
Synonym d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl
PubChem CID 6950340
IUPAC Name (2R)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)C(N)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398965
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L14098.MC
100 mg
£23.20
100mg
Please to purchase this item. Need a web account? Register with us today!
11378965
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L14098.ME
500 mg
£76.30
500mg
Please to purchase this item. Need a web account? Register with us today!
11388965
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L14098.04
2 g
£224.00
2g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

D-tert-Leucine is used for synthesis of amino acids e.g. in the synthesis of aldehydes by double alkylation of unsaturated aldimines of tert-Leu esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-tert-Leucine is used for synthesis of amino acids e.g. in the synthesis of aldehydes by double alkylation of unsaturated aldimines of tert-Leu esters.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

26782-71-8
131.18
NPDBDJFLKKQMCM-UHFFFAOYNA-N
6950340
CC(C)(C)C(N)C(O)=O
C6H13NO2
MFCD00004265
d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl
(2R)-2-amino-3,3-dimethylbutanoic acid

Specifications

>300°C
C6H13NO2
1721825
Soluble in water.
CC(C)(C)C(N)C(O)=O
131.18
131.18
−7° (c=5 in 5N HCl)
26782-71-8
MFCD00004265
d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl
NPDBDJFLKKQMCM-UHFFFAOYNA-N
(2R)-2-amino-3,3-dimethylbutanoic acid
6950340
99%
D-tert-Leucine
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only