Learn More
3M™ Secure Click™ Particulate Filter with Nuisance Level Organic Vapour/Acid Gas Relief
3M™ Secure Click™ Particulate Filter is CE approved for environments containing certain particles. Our filter can also be used for nuisance level organic vapour relief and for ozone protection up to 10 x OEL.
Brand: 3M™ 7100197270
96.70 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.21000kg
Description
- New dual flow cartridges and filters allow for easier breathing
- Protects against particulates with optional nuisance level organic vapour/acid gas relief
- Quick, simple connection. Simply align the connections and push until you hear a click
- Use with 3M™ Secure Click™ Reusable Respirators 800 Series
- Swept-back design allows an enhanced field of view and comfort
Specifications
CE Approved | |
P3R Nuis OV/AG | |
D3138 |
Secure Click™ | |
Filter | |
80/Cs. |