missing translation for 'onlineSavingsMsg'
Learn More
Daidzein, 98%, Thermo Scientific Chemicals Product Code.: 10613552

Daidzein, 98%, Thermo Scientific Chemicals

Product Code. 10613552
25 mg, Glass bottle
Click to view available options
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10613552

Brand: Thermo Scientific Acros 328230250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486-66-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00016954
InChI Key ZQSIJRDFPHDXIC-UHFFFAOYSA-N
Synonym daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyroneShow MoreShow Less
PubChem CID 5281708
ChEBI CHEBI:28197
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 mg
Beilstein 18,II,1
Merck Index 15, 2798
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmso
Formula Weight 254.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Daidzein
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466910 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number328230250,328231000
Lot NumberA0410013 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number328230250,328231000
Lot NumberA0448869 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number328230250,328231000
Lot NumberA0396451 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number328230250,328231000
Lot NumberA0437241 Certificate TypeCertificate of Analysis Date17/12/2021 Catalog Number328230250,328231000
compliance-icons
Product Identifier
  • Daidzein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Daidzein, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.