missing translation for 'onlineSavingsMsg'
Learn More
Genistein, 99%, synthetic, Thermo Scientific Chemicals Product Code.: 10183301

Genistein, 99%, synthetic, Thermo Scientific Chemicals

Product Code. 10183301
25 mg, Glass bottle
Click to view available options
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10183301

Brand: Thermo Scientific Acros 328270250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-72-0
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00016952
InChI Key TZBJGXHYKVUXJN-UHFFFAOYSA-N
Synonym genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genesteinShow MoreShow Less
PubChem CID 5280961
ChEBI CHEBI:28088
IUPAC Name 5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O

Specifications

Melting Point 297.0°C to 298.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 mg
Merck Index 15, 4426
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in dmso and ethanol,soluble in usual organic solvents,soluble in diluted alkalies with yellow color
Formula Weight 270.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Genistein, Synthetic
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463580 Certificate TypeCertificate of Analysis Date10/06/2024 Catalog Number328270250,328271000
Lot NumberA0458236 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog Number328270250,328271000
Lot NumberA0449870 Certificate TypeCertificate of Analysis Date07/02/2023 Catalog Number328270250,328271000
Lot NumberA0370901 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog Number328270250,328271000
Lot NumberA0445656 Certificate TypeCertificate of Analysis Date15/09/2022 Catalog Number328270250,328271000
Product Identifier
  • Genistein (synthetic)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Genistein, 99%, synthetic, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.