missing translation for 'onlineSavingsMsg'
Learn More
Formononetin, 98%, Thermo Scientific Chemicals Product Code.: 10235341

Formononetin, 98%, Thermo Scientific Chemicals

Product Code. 10235341
1 g, Glass bottle
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10235341

Brand: Thermo Scientific Acros 342480010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 485-72-3
Melting Point 256.0°C to 260.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C16H12O4
MDL Number MFCD00016948
Quantity 1 g
Merck Index 15, 4271
Synonym formononetin, biochanin b, formononetol, 7-hydroxy-4'-methoxyisoflavone, 7-hydroxy-3-4-methoxyphenyl-4h-chromen-4-one, 4'-o-methyldaidzein, 7-hydroxy-3-4-methoxyphenyl chromen-4-one, unii-295dqc67bj, 7-hydroxy-3-4-methoxyphenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-methoxyphenyl
InChI Key HKQYGTCOTHHOMP-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C1=COC2=CC(O)=CC=C2C1=O
IUPAC Name 7-hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one
Molecular Weight (g/mol) 268.27
PubChem CID 5280378
ChEBI CHEBI:18088
Formula Weight 268.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Formononetin, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0449059 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number342480010
Lot NumberA0396357 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number342480010
Lot NumberA0408228 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number342480010
Lot NumberA0415378 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number342480010
Lot NumberA0416852 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number342480010
compliance-icons
Product Identifier
  • Formononetin
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formononetin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.