missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Inhibits RNA and DNA synthesis and causes DNA fragmentation in vivo. Reduces tau mRNA levels in vitro.Anticancer agent. Exhibits activity nonlymphocytic leukemia. Identified as targeting human host proteins that interact with SARS-CoV-2.

Specifications
Specifications
CAS | 23541-50-6 |
Molecular Formula | C27H30ClNO10 |
Quantity | 10 mg |
Synonym | daunorubicin hydrochloride, daunorubicin hcl, daunomycin hydrochloride, daunoblastina, rubidomycin hydrochloride, daunorubicinol hydrochloride, daunoblastin, rubomycin, ondena, daunomycin chlorohydrate |
InChI Key | GUGHGUXZJWAIAS-QQYBVWGSSA-N |
SMILES | CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl |
IUPAC Name | (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride |
Molecular Weight (g/mol) | 563.984 |
PubChem CID | 62770 |
Formula Weight | 563.99 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Daunorubicin hydrochloride, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction