missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DCAR/CLEC4B Antibody, R&D Systems™
Goat Polyclonal Antibody
£147.94 - £408.36
Specifications
Antigen | DCAR/CLEC4B |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18798267
|
R&D Systems
AF2757-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18030132
|
R&D Systems
AF2757 |
100 μg |
£408.36
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DCAR/CLEC4B Polyclonal specifically detects DCAR/CLEC4B in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
DCAR/CLEC4B | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Mouse | |
Aplra2, CLEC4B, CLECSF3, LSLCL, P47, SCGF | |
Clec4b1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Q7TS58 | |
6320 | |
Mouse myeloma cell line NS0-derived recombinant mouse DCAR/CLEC4B Gln42-Leu209 Accession # Q7TS58 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse DCAR/CLEC4B in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant mouse DCIR is observed. |
For Research Use Only