missing translation for 'onlineSavingsMsg'
Learn More

Cytiva DEAE Sepharose™ Fast Flow Chromatography Media

For preparative protein separations in both research and industrial applications. Cytiva Life Sciences™ DEAE Sepharose™ Fast Flow Chromatography Media can be used for capture and intermediate purification of wide range of approved biopharmaceuticals.

Brand:  Cytiva 17-0709-10

76.00 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.



Product Code. 10620581

  • £80.00 / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • BioProcess media
  • Weak anion exchanger
  • High chemical stability allows for well proven CIP and sanitization protocols
  • Composed of crosslinked 6% agarose beads with diethylaminoethyl (DEAE)
  • Hydrophilic nature of base matrix ensures low levels of non-specific binding
  • Average particle size of 90μm
  • pH Stability working range from 2 to 12
  • Pack size of 25mL

TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mL
For Preparative Protein Separations in Both Research and Industrial AppliCatIons
DEAE Sepharose Fast Flow Media
45 to 165 μm
15 cm
Weak AnIon Exchanger
6% Cross-Linked Agarose with Dextran surface extenders
20% Ethanol
25 mL
Stable in commonly used aqueous Buffers: 1 M NaOH, 8 M Urea, 8 M Guanidine Hydrochloride, 70% Ethanol
4°C to 30°C
300 to 600 cm/Hoμr
5.9 in.
0.11 to 0.16 mM Cl-/mL
100 kPa
2 to 12
Diethylaminoethyl
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva DEAE Sepharose™ Fast Flow Chromatography Media > DEAE Sepharose Fast Flow chromatography media; Pack size: 25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.