missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decahydronaphthalene, Mixture of cis + trans, 98%, Reagent Grade, Decalin™, Honeywell™
Reagent Grade, 98%
£47.38 - £514.89
Chemical Identifiers
CAS | 91-17-8 |
---|---|
Molecular Formula | C10H18 |
Molecular Weight (g/mol) | 138.254 |
MDL Number | MFCD00004130 |
InChI Key | NNBZCPXTIHJBJL-UHFFFAOYSA-N |
Synonym | decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane |
PubChem CID | 7044 |
ChEBI | CHEBI:38853 |
IUPAC Name | 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene |
SMILES | C1CCC2CCCCC2C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15657810
|
Honeywell Research Chemicals
D251-500ML |
500 mL | Glass bottle | ||||||
15637810
|
Honeywell Research Chemicals
D251-1L |
1 L | Glass bottle | ||||||
15688480
|
Honeywell Research Chemicals
D251-4L |
4 L | Aluminum bottle |
£339.24
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15647810
|
Honeywell Research Chemicals
D251-20L |
20 L | Stainless steel can |
£514.89
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
91-17-8 | |
138.254 | |
NNBZCPXTIHJBJL-UHFFFAOYSA-N | |
7044 | |
1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene |
C10H18 | |
MFCD00004130 | |
decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane | |
CHEBI:38853 | |
C1CCC2CCCCC2C1 |
Specifications
91-17-8 | |
-125°C | |
0.7 to 4.9%, 100°F | |
482°F | |
MFCD00004130 | |
878165 | |
NNBZCPXTIHJBJL-UHFFFAOYSA-N | |
1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene | |
138.254 | |
CHEBI:38853 | |
138.25g/mol | |
Decahydronaphthalene, mixture of cis plus trans |
189° - 191°C | |
0.896g/mL at 25°C | |
57°C (134.6°F) | |
C10H18 | |
UN1147 | |
decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane | |
C1CCC2CCCCC2C1 | |
4.76 (vs air) | |
7044 | |
42mmHg (92°C), 741mmHg (188°C) | |
0.98 |
Safety and Handling
P261-P273-P280-P301 + P310-P305 + P351 + P338-P310
missing translation for 'dotInformation' : class 3 - PG 3 - Decahydronaphthalene
missing translation for 'rtecsNumber' : 202-046-9