Learn More
Decitabine, MedChemExpress
MedChemExpress Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity.
Brand:  Medchem Express HY-A0004
Encompass_Preferred
 View more versions of this product
Product Code. 16428258
  • POA / 10mg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity.
Specifications
Specifications
Decitabine
White
C8H12N4O4
5-Aza-2'-deoxycytidine 5-AZA-CdR NSC 127716
O=C1N=C(N)N=CN1[C@H](O[C@@H]2CO)C[C@@H]2O
228.21
Research
Cancer-programmed cell death
2353-33-5
Research
10 mg
DMSO : ≥ 50 mg/mL (219.10 mM) ∣H2O : 20 mg/mL (87.64 mM; Need ultrasonic)
228.21
95.0%
Solid