missing translation for 'onlineSavingsMsg'
Learn More

Cytiva POM Column Clamp

For use with ÄKTA™ Avant and Pure chromatography systems. Cytiva POM Column Clamp is an acetal plastic (POM) holder that easily inserts into an ÄKTA rail system for holding columns with a 10-21mm O.D.

Brand:  Cytiva 28-9563-19

Product Code. 11310312

  • £60.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pack of five (5) column clamps that hold columns with 10–21mm outer diameter
  • Easily attaches to ÄKTA™ Avant or ÄKTA™ Pure instruments using the rail system:
    • Press inner end tabs of the clamp together
    • Insert pressed end into the rail of the instrument
    • Turn the clamp 90° and release
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clamp
5 Pack
POM
For 10 to 21 mm OD columns
For ÄKTA Avant or ÄKTA Pure Using The Rail System
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva POM Column Clamp > For Use With ÄKTA Avant or ÄKTA Pure Using the Rail System

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.