missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Non-sterile Deep Well Plate for Storage and Collection
Deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
Brand: Corning™
This item is not returnable.
View return policy
Description
- Made of polypropylene
- Offer excellent chemical resistance
- Can withstand temperatures down to -80°C
Specifications
Without Lid | |
Polypropylene | |
Non-sterile | |
1 mL | |
Translucent |
Clear | |
96 | |
Non-treated | |
Round |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction