missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Non-sterile Deep Well Plate for Storage and Collection
Deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
£333.00 - £343.00
Specifications
Lid | Without Lid |
---|---|
Color | Clear |
Material | Polypropylene |
No. of Wells | 96 |
Sterility | Non-sterile |
Product Code | Brand | Well Shape | Total Volume (Metric) Well | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Shape | Total Volume (Metric) Well | Price | Quantity & Availability | ||||
16852250
|
Corning™
3859 |
Round | 1 mL |
£333.00
Pack of 50 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
16862250
|
Corning™
3866 |
Round | 2 mL |
£343.00
Pack of 50 |
Estimated Shipment: 08-11-2024
Log in to see stock. |
||||
16872250
|
Corning™
3947 |
Square, Conical Bottom (V-well) | 2 mL |
£343.00
Pack of 50 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
Description
- Made of polypropylene
- Offer excellent chemical resistance
- Can withstand temperatures down to -80°C
Specifications
Without Lid | |
Polypropylene | |
Non-sterile | |
Translucent |
Clear | |
96 | |
Non-treated |