missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Non-sterile Deep Well Plate for Storage and Collection
Deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
Brand: Corning™ 3947
Additional Details : Weight : 11.22000kg
Description
- Made of polypropylene
- Offer excellent chemical resistance
- Can withstand temperatures down to -80°C
Spécification
Without Lid | |
Polypropylene | |
Non-sterile | |
2 mL | |
Translucent |
Clear | |
96 | |
Non-treated | |
Square, Conical Bottom (V-well) |