Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale
Bench Scales for Basic Industrial Applications
Brand: OHAUS™ 30684745
430.11 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Results are clearly shown using the large, bright backlit LCD display.
- In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
- This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
- Indicator features a stainless steel housing and mounting bracket.
- Features software for Parts Counting, Checkweighing and Totalization.
- Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Specifications
100 Hours with Rechargeable Battery;360 Hours with Disposable Batteries | |
Standard RS232 and optional USB Device or Ethernet | |
150 kg | |
550 x 420 mm | |
D33P150B1L2 | |
Weighing, Checkweighing, Counting, Totalization. | |
17.5 kg |
6 C (LR14) Batteries (Not Included);AC Adapter (Included);Rechargeable Battery (Not Included) | |
ABS plastic indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet | |
7 Segment LCD with Multicolor Backlight | |
20 g | |
-10°C to +40°C, 85%RH, non-condensing | |
686 x 420 x 896 mm |