Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale
Bench Scales for Basic Industrial Applications
£446.00 - £945.00
Specifications
Battery Life | 100 Hours with Rechargeable Battery;360 Hours with Disposable Batteries |
---|---|
Battery Type | 6 C (LR14) Batteries (Not Included);AC Adapter (Included);Rechargeable Battery (Not Included) |
Connection Type | Standard RS232 and optional USB Device or Ethernet |
Construction | ABS plastic indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet |
Display Type | 7-segment Multi-color Backlit LCD |
Product Code | Brand | Capacity | Model | Readability | Dimensions (L x W x H) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Model | Readability | Dimensions (L x W x H) | Price | Quantity & Availability | ||
17512126
|
OHAUS™
30684738 |
15 kg | D33P15B1R1 | 2 g | 489 x 305 x 542 mm |
£515.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17522126
|
OHAUS™
30684739 |
30 kg | D33P30B1R1 | 5 g | 489 x 305 x 542 mm |
£499.20
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17532126
|
OHAUS™
30684740 |
60 kg | D33P60B1R1 | 10 g | 489 x 305 x 542 mm |
£467.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17542126
|
OHAUS™
30684744 |
60 kg | D33P60B1L2 | 10 g | 686 x 420 x 896 mm |
£628.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17552126
|
OHAUS™
30684745 |
150 kg | D33P150B1L2 | 20 g | 686 x 420 x 896 mm |
£555.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17562126
|
OHAUS™
30684748 |
150 kg | D33P150B1X2 | 20 g | 785 x 500 x 896 mm |
£682.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17572126
|
OHAUS™
30684749 |
300 kg | D33P300B1X2 | 50 g | 785 x 500 x 896 mm |
£643.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17582126
|
OHAUS™
30684752 |
300 kg | D33P300B1V3 | 50 g | 933 x 600 x 1195 mm |
£945.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17592126
|
OHAUS™
30684753 |
600 kg | D33P600B1V3 | 100 g | 933 x 600 x 1195 mm |
£891.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17502136
|
OHAUS™
30684756 |
15 kg | D33P15B1R5 | 2 g | 646 x 305 x 130 mm |
£446.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17512136
|
OHAUS™
30684757 |
30 kg | D33P30B1R5 | 5 g | 646 x 305 x 130 mm |
£446.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17522136
|
OHAUS™
30684758 |
60 kg | D33P60B1R5 | 10 g | 646 x 305 x 130 mm |
£446.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17532136
|
OHAUS™
30684762 |
60 kg | D33P60B1L5 | 10 g | 842 x 420 x 134 mm |
£503.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17542136
|
OHAUS™
30684763 |
150 kg | D33P150B1L5 | 20 g | 842 x 420 x 134 mm |
£545.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17552136
|
OHAUS™
30684766 |
150 kg | D33P150B1X5 | 20 g | 941 x 500 x 150 mm |
£731.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17562136
|
OHAUS™
30684767 |
300 kg | D33P300B1X5 | 50 g | 941 x 500 x 150 mm |
£731.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Results are clearly shown using the large, bright backlit LCD display.
- In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
- This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
- Indicator features a stainless steel housing and mounting bracket.
- Features software for Parts Counting, Checkweighing and Totalization.
- Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Specifications
100 Hours with Rechargeable Battery;360 Hours with Disposable Batteries | |
Standard RS232 and optional USB Device or Ethernet | |
7-segment Multi-color Backlit LCD | |
Weighing, Checkweighing, Counting, Totalization. |
6 C (LR14) Batteries (Not Included);AC Adapter (Included);Rechargeable Battery (Not Included) | |
ABS plastic indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet | |
-10°C to +40°C, 85%RH, non-condensing |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
OHAUS™ Defender™ 3000 - I-D33 Bench Scale