Learn More
OHAUS™ Defender™ 3000 Washdown - I-D33 Bench Scale (GB)
Stainless Steel Bench Scales for Basic Industrial Washdown Applications
Brand: OHAUS™ 30685086
Additional Details : Weight : 19.21000kg Transport : UN number : 2800 Chem class : 8
Description
- Results are clearly shown using the large, bright backlit LCD display.
- In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
- This industrial scale features a 304 stainless steel platform and frame, IP67 load cell.
- Indicator features an IP66 washdown protected 304-stainless steel housing with mounting bracket.
- Features software for Parts Counting, Checkweighing and Totalization.
- Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Specifications
100 Hours with Rechargeable Battery | |
Standard RS232 and optional USB Device or Ethernet | |
60 kg | |
550 x 420 mm | |
D33XW60C1L7 | |
-10°C to +40°C, 85%RH, non-condensing | |
688 x 420 x 922 mm |
AC Adapter (Included);Rechargeable Battery (Included) | |
304-grade stainless steel platform and frame, stainless steel IP66 indicator, IP67 load cell, non-slip adjustable rubber feet | |
7 Segment LCD with Multicolor Backlight | |
10 g | |
IP66 | |
Weighing, Checkweighing, Counting, Totalization. | |
19.2 kg |