Learn More
OHAUS™ Defender™ 5000 - D52 Bench Scale, Models TD52XW RQL
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
£1220.00 - £1330.00
Specifications
Battery Life 20 Hours with Rechargeable Battery
Connection Type Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Construction Stainless steel (TD52XW) indicator, 304-grade stainless steel platform with sand gray powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet
Display Type Backlit LCD
Temperature (Metric) Operating -10°C to +40°C
View More Specs
Products 6
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
15968525
View Documents
OHAUS™
30444833
D52XW15RQDL2-GB
£1220.00
1 unit
15968585
View Documents
OHAUS™
30461581
D52XW15RQDL2-M
£1330.00
1 unit
15978525
View Documents Promotion Details
OHAUS™
30444834
D52XW30RQDL2-GB
£1325.00
1 unit
15978585
View Documents
OHAUS™
30461582
D52XW30RQDL2-M
£1330.00
1 unit
15988525
View Documents
OHAUS™
30444835
D52XW60RQDL2-GB
£1220.00
1 unit
15988585
View Documents
OHAUS™
30461583
D52XW60RQDL2-M
£1330.00
1 unit
Description
Description
  • User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
  • Durable construction with a stainless steel platform, sand gray powder-coated steel frame and aluminum load cell.
  • Defender meets NTEP/Measurement Canada or OIML/EC type approvals (depending on region).
  • Features common industrial modes and supports a 50000-item library with 1000 user profiles for easy data management.
  • Includes GMP/GLP with time/date for result traceability.
Specifications
Specifications
20 Hours with Rechargeable Battery
Stainless steel (TD52XW) indicator, 304-grade stainless steel platform with sand gray powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet
-10°C to +40°C
18.200 kg
Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Backlit LCD
570 x 400 x 934 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
OHAUS™ Defender™ 5000 - D52 Bench Scale, Models TD52XW RQL