Learn More
Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Brand:  Thermo Scientific™ D0836
189.56 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 10297093
  • £209.09 / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cartridge for pretreatment macropure; macroreticular, activated carbon; full size
Macroreticular, Activated Carbon
B-Pure™
2000 gal.
Pretreatment Macropure
Deionizer Cartridge
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Deionizer Cartridges > Cartridge for pretreatment macropure; macroreticular, activated carbon; full size