Learn More
Delgocitinib, MedChemExpress
MedChemExpress Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.
Brand: Medchem Express HY-109053
This item is not returnable.
View return policy
Description
Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.Specifications
Delgocitinib | |
Off-White | |
C16H18N6O | |
JTE-052 | |
O=C(N(C[C@@H]1C)[C@]21CN(C3=C(C=CN4)C4=NC=N3)CC2)CC#N | |
310.35 | |
Research | |
COVID-19-immunoregulation |
1263774-59-9 | |
Research | |
1 mg | |
DMSO : ≥ 58 mg/mL (186.89 mM) | |
310.35 | |
99.75% | |
Solid |