missing translation for 'onlineSavingsMsg'
Learn More

Delgocitinib, MedChemExpress

MedChemExpress Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.

£240.79 - £6293.21

Specifications

Chemical Name or Material Delgocitinib
CAS 1263774-59-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H18N6O
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16450597
Encompass_Preferred
View Documents
Medchem Express
HY-109053
50 mg
£4005.10
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16446987
Encompass_Preferred
View Documents
Medchem Express
HY-109053
5 mg
£744.15
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16445196
Encompass_Preferred
View Documents
Medchem Express
HY-109053
100 mg
£6293.21
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16443016
Encompass_Preferred
View Documents
Medchem Express
HY-109053
25 mg
£2518.01
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16434268
Encompass_Preferred
View Documents
Medchem Express
HY-109053
10 mg
£1258.40
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16421129
Encompass_Preferred
View Documents
Medchem Express
HY-109053
1 mg
£240.79
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.
Specifications

Specifications

Delgocitinib
Off-White
C16H18N6O
DMSO : ≥ 58 mg/mL (186.89 mM)
310.35
99.75%
Solid
1263774-59-9
Research
JTE-052
O=C(N(C[C@@H]1C)[C@]21CN(C3=C(C=CN4)C4=NC=N3)CC2)CC#N
310.35
Research
COVID-19-immunoregulation
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.