Learn More
Emma Safety Footwear Dempo Safety Shoes
Extreme safety in extreme temperatures. Easy on/off slip-on lightweight leather boot with a double density rubber-PU sole. The Nitrile rubber outsole repels corrosive chemicals and substances, even liquid, molten metals. Heat resistant up to 300°C.
Brand:  Emma Safety Footwear MM58384846
 View more versions of this product
Product Code. 15918801
  • £193.00 / 1 pair
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
Dempo
Leather
Steel
Black
Hydro-Tec™ Sanitized Silver lined tongue
46