Learn More
Emma Safety Footwear Dempo Safety Shoes
Extreme safety in extreme temperatures. Easy on/off slip-on lightweight leather boot with a double density rubber-PU sole. The Nitrile rubber outsole repels corrosive chemicals and substances, even liquid, molten metals. Heat resistant up to 300°C.
£180.20 - £181.26
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Color | Black |
Design | Dempo |
Liner Material | Hydro-Tec™ Sanitized Silver lined tongue |
Material | Leather |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15928791
|
Emma Safety Footwear
MM58384837 |
37 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15938791
|
Emma Safety Footwear
MM58384838 |
38 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15948791
|
Emma Safety Footwear
MM58384839 |
39 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15958791
|
Emma Safety Footwear
MM58384840 |
40 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15968791
|
Emma Safety Footwear
MM58384841 |
41 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15978791
|
Emma Safety Footwear
MM58384842 |
42 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15988791
|
Emma Safety Footwear
MM58384843 |
43 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15998791
|
Emma Safety Footwear
MM58384844 |
44 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15908801
|
Emma Safety Footwear
MM58384845 |
45 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15918801
|
Emma Safety Footwear
MM58384846 |
46 |
£180.20
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15928801
|
Emma Safety Footwear
MM58384847 |
47 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15938801
|
Emma Safety Footwear
MM58384848 |
48 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15948801
|
Emma Safety Footwear
MM58384849 |
49 |
£181.26
1 pair |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Sanitized
Specifications
EN ISO 20345:2011 | |
Dempo | |
Leather |
Black | |
Hydro-Tec™ Sanitized Silver lined tongue | |
Steel |