Learn More
Emma Safety Footwear Dempo Safety Shoes
Extreme safety in extreme temperatures. Easy on/off slip-on lightweight leather boot with a double density rubber-PU sole. The Nitrile rubber outsole repels corrosive chemicals and substances, even liquid, molten metals. Heat resistant up to 300°C.
Brand: Emma Safety Footwear MM58384841
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Sanitized
Specifications
EN ISO 20345:2011 | |
Dempo | |
Leather | |
Steel |
Black | |
Hydro-Tec™ Sanitized Silver lined tongue | |
41 |