Learn More
BRAND™ Desiccator with Knob Lid
Desiccator with Knob Lid includes a studded base ring for improved stability.
£195.00 - £250.00
Specifications
Material Borosilicate Glass 3.3
Product Type Desiccator
Includes Knob Lid
View More Specs
Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
10728051
View Documents
BRAND™
65031
150 mm
£195.00
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10470673
View Documents
BRAND™
65038
200 mm
£250.00
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Borosilicate Glass 3.3
  • DIN 12491
  • Precisely ground flat flanges
  • Base ring surface studded to improve stability and help prevent cracks
  • Interchangeable lid
Specifications
Specifications
Borosilicate Glass 3.3
Desiccator
Knob Lid
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BRAND™ Desiccator with Knob Lid