missing translation for 'onlineSavingsMsg'
Learn More

SymLine™ Desktop and Wall Mounts

The filling units are mounted in space-saving fashion within your workspace. Available as a special HPLC connection, for direct connection to your waste tubing.

Brand:  SymLine™ 106601

298.82 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 17327419

  • £372.00 / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Desktop mount can be integrated into nearly all working areas
  • Whether used in conjunction with a work table or a fume cupboard, it always provides efficient transfer of liquid waste into connected waste systems
  • Flat desktop mount and HPLC waste collector in one
  • Integrated connections for capillaries and tubes allow this installation to protrude only slightly from the worktop
  • With the Rear Wall Mount, you use the available capacity of your working area for even more space for laboratory samples and experimental setups
  • You can dispose waste directly in the fume cupboard
Specifications

Specifications

GL 45 (m), Lower Thread GL 25 (m)
Transfer of Liquids Into Connected Waste Systems
HDPE-EC
Mount with Mounting Flange, Long Shaft for XL-funnel, two 90° Bends
3 x and 4 x Collector, SafetyWasteCap, Safety Funnel and Pipe System
Wall Mount
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers