Learn More
SymLine™ Desktop and Wall Mounts
The filling units are mounted in space-saving fashion within your workspace. Available as a special HPLC connection, for direct connection to your waste tubing.
Brand: SymLine™ 106669
Additional Details : Weight : 0.10200kg
Description
- Desktop mount can be integrated into nearly all working areas
- Whether used in conjunction with a work table or a fume cupboard, it always provides efficient transfer of liquid waste into connected waste systems
- Flat desktop mount and HPLC waste collector in one
- Integrated connections for capillaries and tubes allow this installation to protrude only slightly from the worktop
- With the Rear Wall Mount, you use the available capacity of your working area for even more space for laboratory samples and experimental setups
- You can dispose waste directly in the fume cupboard
Specifications
4 x UNF 1/4 in. 28G | |
Fittings for Capillary Dia. 2.3/3.2 mm, 2 x NPT 1/8 in., Tube Connector for Dia. 5 to 11.5 mm, Pad Dia. 53 mm, 32 mm OD Pipe | |
3 x and 4 x Collector, SafetyWasteCap, Safety Funnel and Pipe System | |
Desktop Mount |
Black | |
Transfer of Liquids Into Connected Waste Systems | |
HDPE |