Learn More
DWK Life Sciences Porcelain Desiccator Plate
The desiccator insert is a sieve plate on which the substance to be dried is placed in an appropriate container.
Brand: DWK Life Sciences 297256103
101.63 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The desiccator insert is a sieve plate that separates the desiccator base into two areas - there is a desiccant in the lower part and the substance to be dried is placed in the upper part. The substance to be dried is placed on the sieve plate in an appropriate container for drying.
- The heat-resistant material allows hot samples to be placed into the desiccator
- Made of heat-resistant material
Specifications
190 mm | |
Porcelain | |
Drying of materials |