missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Porcelain Desiccator Plate
The desiccator insert is a sieve plate on which the substance to be dried is placed in an appropriate container.
£32.05 - £184.00
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
10409542
|
DWK Life Sciences
297255707 |
140 mm |
£49.80
Each |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
10325011
|
DWK Life Sciences
297256103 |
190 mm |
£96.75
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
10672062
|
DWK Life Sciences
297256909 |
280 mm |
£184.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
10099820
|
DWK Life Sciences
297256609 |
235 mm |
£133.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
10020091
|
DWK Life Sciences
297254608 |
90 mm |
£32.05
Each |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
Description
The desiccator insert is a sieve plate that separates the desiccator base into two areas - there is a desiccant in the lower part and the substance to be dried is placed in the upper part. The substance to be dried is placed on the sieve plate in an appropriate container for drying.
- The heat-resistant material allows hot samples to be placed into the desiccator
- Made of heat-resistant material
Specifications
Porcelain | |
Drying of materials |