Learn More
DWK Life Sciences Porcelain Desiccator Plate
The desiccator insert is a sieve plate on which the substance to be dried is placed in an appropriate container.
Brand:  DWK Life Sciences 297255707
Code : NEW
Additional Details : Weight : 0.28500kg
 View more versions of this product
Product Code. 10409542
  • £49.80 / Each
Estimated Shipment: 03-10-2024
to see stock.
Add to basket
Description
Description
The desiccator insert is a sieve plate that separates the desiccator base into two areas - there is a desiccant in the lower part and the substance to be dried is placed in the upper part. The substance to be dried is placed on the sieve plate in an appropriate container for drying.
  • The heat-resistant material allows hot samples to be placed into the desiccator
  • Made of heat-resistant material
Specifications
Specifications
140 mm
Porcelain
Drying of materials