Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand: Fisherbrand™ 11957-78
Description
- Produced in state of the art manufacturing facilities approved by lamp manufacturers
- Full traceability of manufactured products
- Lamps available for all major instruments
Specifications
UV-A, UV-C | |
0.5 mm | |
1 Ea. |
400/185 nm | |
Waters 996, 2996 PDA long Life | |
Deuterium Lamp |